missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL1 Monoclonal Antibody (OTI3G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810642
This item is not returnable.
View return policy
Description
Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in fast skeletal muscle. Two transcript variants have been identified for this gene.Specifications
| MYL1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P05976 | |
| MYL1 | |
| Human recombit protein fragment corresponding to amino acids 2-150 of human MYL1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3G8 | |
| Unconjugated | |
| MYL1 | |
| MLC1F, MLC3F | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4632 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction