missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MYL1 Polyclonal Antibody
GREENER_CHOICE

Product Code. p-200128861
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17720704 200 μL 200µL
17793144 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17720704 Supplier Invitrogen™ Supplier No. PA5119296

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MYL1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in fast skeletal muscle. Two transcript variants have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene MYL1
Gene Accession No. P02600, P05977
Gene Alias A1 catalytic; A2 catalytic; AI325107; alkali myosin light chain 1; alkali myosin light chain 3; alkali myosin light chain 3 A2 catalytic; Ca2+-binding protein-like protein; fast myosin alkali light chain 1; fast myosin alkali light chain 3; fast myosin light chain 1; fast myosin light chain 1F; Mcl3; MLC; Mlc1; MLC1/MLC3; MLC1F; MLC1F/MLC3F; Mlc3; MLC3F; MLC3-f; MLCf; Myl1; Mylf; myosin light chain; myosin light chain 1; myosin light chain 1, skeletal muscle isoform; myosin light chain 1/3, skeletal muscle isoform; myosin light chain 3; myosin light chain 3, skeletal muscle isoform; myosin light chain A1/A2; Myosin light chain alkali 1/2; myosin light chain, alkali, fast skeletal muscle; myosin, light chain 1; myosin, light chain 1, alkali; skeletal, fast; myosin, light polypeptide 1; myosin, light polypeptide 1, alkali; atrial, embryonic; myosin, light polypeptide 1, alkali; skeletal, fast; myosin, light polypeptide 4, alkali; atrial, embryonic
Gene Symbols MYL1
Host Species Rabbit
Immunogen Recombinant protein Myosin Light Chain 1. The antigen corresponds to amino acid range 3-139 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17901, 56781
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.