missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL4 Antibody (4E8), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42646AF647
This item is not returnable.
View return policy
Description
MYL4 Monoclonal specifically detects MYL4 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| MYL4 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MYL4 | |
| Recombinant human MYL4 (1-197aa) purified from E. coli | |
| 100 μL | |
| Vision | |
| 4635 | |
| Store at 4C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 400000000 | |
| Western Blot, ELISA | |
| ALC1, AMLC, GT1, MLC1, Myosin light chain 1, embryonic muscle/atrial isoform, myosin light chain 4, Myosin light chain alkali GT-1 isoform, myosin, atrial/fetal muscle, light chain, myosin, light chain 4, alkali; atrial, embryonic, myosin, light polypeptide 4, alkali; atrial, embryonic, PRO1957 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction