missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL4 Monoclonal Antibody (OTI3H6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807580S
This item is not returnable.
View return policy
Description
MYL4 Monoclonal antibody specifically detects MYL4 in Human samples. It is validated for Western BlotSpecifications
| MYL4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P12829 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4635 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3H6 | |
| Unconjugated | |
| MYL4 | |
| ALC1; AMLC; ELC; ELC1a; GT1; MLC1; MLC1a; MLC1EMB; MYL4; Myla; myosin light chain 1, atrial isoform; myosin light chain 1, atrial/fetal isoform; myosin light chain 1, embryonic muscle/atrial isoform; Myosin light chain 4; myosin light chain alkali GT-1 isoform; myosin light chain, alkali, cardiac atria; myosin, atrial/fetal muscle, light chain; myosin, light chain 4; myosin, light chain 4, alkali; atrial, embryonic; myosin, light polypeptide 4; myosin, light polypeptide 4, alkali; atrial, embryonic; PRO1957 | |
| Human recombinant protein fragment corresponding to amino acids 2-197 of human MYL4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction