missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYL5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557928
This item is not returnable.
View return policy
Description
Immunogen sequence: NAFKMLDPDG KGKINKEYIK RLLMSQADKM TAEEVDQMFQ FASIDVAGNL DYKALSYVI Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 53%.
This gene encodes one of the myosin light chains, a component of the hexameric ATPase cellular motor protein myosin. Myosin is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene product, one of the regulatory light chains, is expressed in fetal muscle and in adult retina, cerebellum, and basal ganglia.
Specifications
| MYL5 | |
| Polyclonal | |
| Unconjugated | |
| MYL5 | |
| MYL5; MYLC2; myLC-2; Myosin light chain 5; myosin regulatory light chain 5; myosin, light chain 5, regulatory; myosin, light polypeptide 5, regulatory; superfast myosin regulatory light chain 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4636 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q02045 | |
| MYL5 | |
| Recombinant protein corresponding to Human MYL5. Recombinant protein control fragment (Product #RP-96761). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction