missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-30441-25ul
This item is not returnable.
View return policy
Description
MYL6 Polyclonal specifically detects MYL6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MYL6 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| P60660 | |
| MYL6 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MTEEEVEMLVAGHEDSNGCINYEAF | |
| 25ul | |
| Vision | |
| 4637 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ESMLC, LC17, LC17A, LC17B, LC17-GI, LC17-NM, MLC1SM, MLC-3, MLC3NM, MLC3SM, Myosin light chain 3, Myosin light chain A3, Myosin light chain alkali 3, myosin light polypeptide 6,17 kDa myosin light chain, myosin, light chain 6, alkali, smooth muscle and non-muscle, myosin, light polypeptide 6, alkali, smooth muscle and non-muscle, Smooth muscle and nonmuscle myosin light chain alkali 6 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction