missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19449
This item is not returnable.
View return policy
Description
MYL7 Polyclonal specifically detects MYL7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MYL7 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) with 0.025% Proclin 300 | |
| atrial isoform, light polypeptide 7, regulatory, MLC-2a, MYL2A, MYLC2AMLC2a, myosin, light chain 7, regulatory | |
| Rabbit | |
| 19 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse. Species reactivity based on sequence identity: Rat (94%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:150000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q01449 | |
| MYL7 | |
| Recombinant protein encompassing a sequence within the center region of human MYL7. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 58498 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction