missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL7 Antibody (OTI4E7), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72859V
This item is not returnable.
View return policy
Description
MYL7 Monoclonal specifically detects MYL7 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| MYL7 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MYL7 | |
| Full length recombinant protein of human MYL7 (NP_067046) produced in E.coli. | |
| 0.1 mL | |
| 58498.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI4E7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| atrial isoform, light polypeptide 7, regulatory, MLC-2a, MYL2A, MYLC2AMLC2a, myosin, light chain 7, regulatory | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction