Learn More
MYL7 Mouse anti-Human, Clone: OTI3A8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF800218
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Encoded by the MYL7 gene, myosin regulatory light chain 7, also designated Myosin regulatory light chain 2, atrial isoform (MLC-2a), is part of a hexameric complex of two heavy chains and four light chains predomitly expressed in adult atrial muscle. Myosin regulatory light chain 7 binds calcium and has been shown to be a useful molecular marker for cardiac chamber specification. The co-expression of myosin regulatory light chain 7 and myosin light chain 2 (MLC2v) in the outflow tract and atrioventricular canal, together with the single expression in the atrial (MYL7) and ventricular (MYL2) myo-cardium, permits the delineation of their boundaries. At the amino acid level there is 95% homology between the human and mouse myosin regulatory light chain 7 sequences.Specifications
| MYL7 | |
| Monoclonal | |
| Unconjugated | |
| MYL7 | |
| MYL2A, MYLC2A | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 58498 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3A8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q01449 | |
| MYL7 | |
| Full length human recombit protein of human MYL7 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.