Learn More
MYL9 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-15354-100UL
Description
The MYL9 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MYL9. This antibody recognizes human antigen. The MYL9 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| MYL9 | |
| 1000 μg/mL | |
| 20 kDa myosin light chain, LC20, MLC 2C, MLC2, MRLC1, MYL9, Myosin RLC, MYRL2 | |
| 100 μL | |
| 10398 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |