missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYLIP Mouse anti-Human, Clone: OTI3A2, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807457S
This item is not returnable.
View return policy
Description
MYLIP Monoclonal antibody specifically detects MYLIP in Human samples. It is validated for Western BlotSpecifications
| MYLIP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WY64 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 29116 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI3A2 | |
| Unconjugated | |
| MYLIP | |
| 9430057C20Rik; band 4.1 superfamily member BZF1; BM-023; BZF1; cellular modulator of immune recognition (c-MIR); E3 ubiquitin ligase-inducible degrader of the low density lipoprotein receptor; E3 ubiquitin-protein ligase MYLIP; I79_001311; IDOL; inducible degrader of the LDL-receptor; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase MYLIP; MIR; Mylip; Myosin regulatory light chain interacting protein; myosin regulatory light chain-interacting protein; PP5242; RING-type E3 ubiquitin transferase MYLIP | |
| Human recombinant protein fragment corresponding to amino acids 127-385 of human MYLIP produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction