missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYLK4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98058-50ul
This item is not returnable.
View return policy
Description
MYLK4 Polyclonal antibody specifically detects MYLK4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MYLK4 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| caMLCK like, EC 2.7.11, EC 2.7.11.1, myosin light chain kinase family, member 4, SGK085, SgK085myosin light chain kinase family member 4, Sugen kinase 85 | |
| Produced in rabbits immunized with E. coli-derived Human MYLK4 fragment. | |
| 50 μg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 340156 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction