missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYLPF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536041
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 19 kDa. Purity is >95% by SDS-PAGE.
MYLPF (myosin light chain, phosphorylatable, fast skeletal muscle), also known as fast skeletal myosin light chain 2 or MLC2B, is a 169 amino acid protein that is expressed in fetal and adult skeletal muscle. A calcium binding protein, MYLPF contains three EF hand domains and is encoded by a gene that maps to human chromosome 16p11.2. Chromosome 16 encodes over 900 genes in approximately 90 million base pairs, makes up nearly 3% of human cellular DNA and is associated with a variety of genetic disorders. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth.
Specifications
| MYLPF | |
| Polyclonal | |
| Unconjugated | |
| MYLPF | |
| 2410014J02Rik; DKFZp779C0757; DTNB; fast skeletal myosin light chain 2; G2; HUMMLC2B; MGC13450; MLC2; MLC-2; MLC2B; MLC2F; MRLC2; MYL11; Myl2; MYLPF; Myolc1; myosin light chain; myosin light chain 2; myosin light chain 2 type 2; myosin light chain 2 type I; myosin light chain, phosphorylatable, fast skeletal muscle; Myosin light polypeptide 2 alkali; myosin regulatory light chain 2, skeletal muscle isoform; Myosin regulatory light chain 2, skeletal muscle isoform type 2; myosin, light chain 11, regulatory; Myosin, light polypeptide 2, alkali; Myosin, light polypeptide 2, alkali; ventricular, skeletal, slow; phosphorylatable fast skeletal muscle myosin light chain; Unknown (protein for MGC:143425); ventricular skeletal slow; ventricular, skeletal, slow | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17907, 24584, 29895 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P04466, P97457, Q96A32 | |
| MYLPF | |
| Synthetic peptide corresponding to the N-terminal of human MYLPF. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction