Learn More
MYNN Mouse anti-Human, Clone: OTI1F10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF800192
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the BTB/POZ and zinc finger domain-containing protein family that are involved in the control of gene expression. Alternative splicing results in multiple transcript variants and a pseudogene has been identified on chromosome 14.Specifications
| MYNN | |
| Monoclonal | |
| Unconjugated | |
| MYNN | |
| OSZF, SBBIZ1, ZBTB31, ZNF902 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55892 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1F10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9NPC7 | |
| MYNN | |
| Human recombit protein fragment corresponding to amino acids 10-300 of human MYNN produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.