missing translation for 'onlineSavingsMsg'
Learn More

MYO18A Antibody, DyLight 755, Novus Biologicals™

Product Code. 18718302 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18718302 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18718302 Supplier Novus Biologicals Supplier No. NB10079840IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MYO18A Polyclonal antibody specifically detects MYO18A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO18A
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KIAA0216DKFZp686L0243, MAJN, Molecule associated with JAK3 N-terminus, myosin 18A, Myosin containing a PDZ domain, myosin containing PDZ domain, myosin XVIIIA, myosin-XVIIIa, MYSPDZ, SP-A receptor subunit SP-R210 alphaS, SPR210
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 2004 andd 2054 of human myosin 18A using the numbering given in entry NP_510880.2 (GeneID 399687).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 399687
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.