missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MYO19 Antibody (SR4212), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27822-50ul
This item is not returnable.
View return policy
Description
MYO19 Monoclonal antibody specifically detects MYO19 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| MYO19 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 80179 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR4212 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| FLJ22865, MYOHD1, myosin head domain containing 1, Myosin head domain-containing protein 1, myosin XIX, myosin-XIX | |
| A synthesized peptide derived from human MYO19 (Uniprot #: Q96H55) | |
| 50 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction