missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYO19 Recombinant Rabbit Monoclonal Antibody (JE64-30)

MYO19 Recombinant Rabbit Monoclonal Antibody (JE64-30)
Brand: Invitrogen MA544757
This item is not returnable.
View return policy
Description
MYO19 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Specifications
| MYO19 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q96H55 | |
| MYO19 | |
| Recombinant protein within human MYO19 aa 431-680/970. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| JE64-30 | |
| Unconjugated | |
| MYO19 | |
| 1110055A02Rik; MYO19; MYOHD1; myosin head domain containing 1; myosin head domain-containing protein 1; myosin XIX; myosin-XIX; unconventional myosin-XIX | |
| Rabbit | |
| Protein A | |
| RUO | |
| 80179 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction