missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYO1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563820
This item is not returnable.
View return policy
Description
Immunogen sequence: EVVNKINRAN GKSTSRIFLL TNNNLLLADQ KSGQIKSEVP LVDVTKVSMS SQNDGFFAVH LKEGSEAASK GDFLFSSDHL IEMATKLYRT TLSQTKQKLN IEISDEFLVQ FR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Motor protein that may participate in process critical to neuronal development and function such as cell migration, neurite outgrowth and vesicular transport.
Specifications
| MYO1B | |
| Polyclonal | |
| Unconjugated | |
| MYO1B | |
| AA406997; MIH-L; MMIa; MMI-alpha; molecular motor; MYH-1c; Myo1a; MYO1B; MYO1B variant protein; myosin heavy chain myr 1; Myosin I alpha; Myosin IA (Myosin I alpha) (MMI-alpha) (MMIa) (MIH-L); myosin Ib; myosin-I alpha; myosin-Ib; myr1; Unconventional myosin-Ib | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4430 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43795 | |
| MYO1B | |
| Recombinant protein corresponding to Human MYO1B. Recombinant protein control fragment (Product #RP-103368). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction