Learn More
MYO1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15012-1-AP-150UL
This item is not returnable.
View return policy
Description
Motor protein that may participate in process critical to neuronal development and function such as cell migration, neurite outgrowth and vesicular transport.Specifications
| MYO1B | |
| Polyclonal | |
| Unconjugated | |
| MYO1B | |
| MMI alpha, MMIa, MYH 1c, MYO1B, Myosin I alpha, myosin IB, myr1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117057, 17912, 4430 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43795, P46735, Q05096 | |
| Myo1b | |
| MYO1B Fusion Protein Ag6895 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido