missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYO1C Polyclonal Antibody

Description
Immunogen sequence: GEEETLRRLG LERNPQSYLY LVKGQCAKVS SINDKSDWKV VRKALTVIDF TEDEVEDLLS IVASVLHLGN IHFAANEESN AQVTTENQLK YLTRLLSVEG STLREALTHR KIIAKGEELL SPLNLEQAAY ARDALAKAVY SRTFTWLV Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.
MYO1C is a member of the unconventional myosin protein family, which are actin-based molecular motors. The protein is found in the cytoplasm, and one isoform with a unique N-terminus is also found in the nucleus. The nuclear isoform associates with RNA polymerase I and II and functions in transcription initiation. The mouse ortholog of this protein also functions in intracellular vesicle transport to the plasma membrane.
Specifications
Specifications
| Antigen | MYO1C |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MYO1C |
| Gene Accession No. | O00159 |
| Gene Alias | C80397; FLJ23903; mm1beta; MMIb; MMI-beta; myo1c; myo1ca; MYO1E; myosin 1c, paralog a; myosin heavy chain myr 2; myosin I; Myosin I beta; myosin IB; myosin IC; myosin Ic, paralog a; myosin IC-like; myosin-I beta; myosin-Ibeta; myosin-Ic; Myr2; NMI; nuclear myosin I; nuclear myosin I beta; si:ch211-69i14.4; unconventional myosin Myr2 I heavy chain; unconventional myosin-Ic |
| Gene Symbols | MYO1C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?