Learn More
MYO1G Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25772-1-AP-20UL
 View more versions of this product
Product Code. 18019395
  • € 145.00 / 20µL
Description
The MYO1G antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MYO1G. This antibody recognizes human antigen. The MYO1G antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
MYO1G
550 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
HA 2, MYO1G, myosin IG
20 μL
64005
Store at -20°C. Stable for one year after shipment.