Learn More
MYO1G Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25772-1-AP-150UL
This item is not returnable.
View return policy
Description
The MYO1G antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MYO1G. This antibody recognizes human antigen. The MYO1G antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| MYO1G | |
| 550 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| HA 2, MYO1G, myosin IG | |
| 150 μL | |
| 64005 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |