missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYO1H Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558825
This item is not returnable.
View return policy
Description
Immunogen sequence: VSTSNLSDGI LVIHVSPEDS KQKGDAVLQC GHVFEAVTKL VMLVKKENIV NVVQGSLQFF ISPGKEGTIV FDTGPEEQVY KNKNGQLTVV SG Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 72%.
Specifications
| MYO1H | |
| Polyclonal | |
| Unconjugated | |
| MYO1H | |
| 4631401O15Rik; MYO1H; myosin 1H; myosin IH; myosin-1H; myosin-Ih; unconventional myosin-Ih | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 283446 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N1T3 | |
| MYO1H | |
| Recombinant protein corresponding to Human MYO1H. Recombinant protein control fragment (Product #RP-97851). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction