missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MYO3A (MYO3 alpha), GST Tag Recombinant Protein

Product Code. 17118755
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17118755 100 μg 100µg
15990303 10 μg 10µg
17188795 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17118755 Supplier Invitrogen™ Supplier No. A32899

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This MYO3A (MYO3 alpha) recombinant human protein (1-434) was expressed in insect cells. MYO3A (MYO3 alpha) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Catalytic The gene accession number for MYO3A (MYO3 alpha) is NP_059129.3.

MYO3A (MYO3 alpha) serine/threonine kinase belongs to the myosin superfamily where they are categorized into conventional myosins (class II) and unconventional myosins (classes I and III through XV) based on their variable C-terminal cargo-binding domains. Class III myosins, such as MYO3A, have a kinase domain N-terminal to the conserved N-terminal motor domains and are expressed in photoreceptors. MYO3A plays an important role in hearing in humans and mutations in the MYO3A have been shown to cause nonsyndromic progressive hearing loss. MYO3A is highly expressed in retina and cochlea.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NEV4
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.25mM DTT, 0.1mM PMSF, 0.1mM EDTA, 10mM glutathione, 150mM NaCl, 25% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 53904
Molecular Weight (g/mol) 75.3 kDa
Name Human MYO3A (MYO3 alpha), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias DFNB30; MYO3A; myosin IIIA; myosin-IIIa
Common Name MYO3A
Gene Symbol MYO3A
KinaseFamily STE20 Kinase Family
Protein Length 1-434
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 1-434
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Recombinant, Active, Catalytic
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.