missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYO3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561016
This item is not returnable.
View return policy
Description
Immunogen sequence: FISQCLIKDF ERRPSVTHLL DHPFIKGVHG KVLFLQKQLA KVLQDQKHQN PVAKTRHERM HTRRPYHVED AEKYCLEDDL VNLEVLDEDT IIHQLQKRYA DLLIY Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.
MYO3B (MYO3 beta) is a member of the Class III myosins that are actin-dependent motor proteins containing an amino-terminal kinase domain. MYO3B contains a N-terminal kinase domain, followed by motor, neck, and tail domains. The MYO3B gene generates a variety of splice variants that contain 1 or 2 calmodulin-binding (IQ) motifs in the neck domain and 1 of 3 domains in the tail domain. Northern blot analysis shows expression of a 7-kb MYO3B transcript in the human retina but not in a RPE cell line. The MYO3B gene transcript is also detected in the kidney, intestine and testis. The MYO3B gene maps to chromosome 2q31.1-q31.2 by genomic sequence alignment.
Specifications
| MYO3B | |
| Polyclonal | |
| Unconjugated | |
| MYO3B | |
| A430065P19Rik; MYO3B; myosin IIIB; myosin IIIB1; myosin IIIB2; myosin-IIIb; RGD1560313 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140469 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WXR4 | |
| MYO3B | |
| Recombinant protein corresponding to Human MYO3B. Recombinant protein control fragment (Product #RP-102978). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction