Learn More
Invitrogen™ MYO6 Monoclonal Antibody (K70041_12A6), Invitrogen™
Mouse Monoclonal Antibody
€ 524.00 - € 876.00
Specifications
| Antigen | MYO6 |
|---|---|
| Clone | K70041_12A6 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30204931
|
Invitrogen™
MA555888 |
50 μL |
€ 524.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30212649
|
Invitrogen™
MA556268 |
200 μL |
€ 876.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: EEAERLRRIQ EEMEKERKRR EEDEKRRRKE EEERRMKLEM EAKRKQEEEE RKKREDDEKR IQAEVEAQLA RQKEEESQQQ AVLEQERRDR ELALRIAQSE AELISDEAQA DLALRRSLDS YPVSKNDGTR PKMTPEQMAK EMSEFLSRGP AVLATKAAAG TKKYDLSKWK YAELRDTINT SCDIELLAAC REEFHRRLKV YHAWKSKNKK RNTETEQRAP KSVTDYDFAP FLNNSPQQNP AAQIPARQRE IEMNRQQRFF RIPFIRPADQ YKDPQSKKKG WWYAHFDGPW IARQMELHPD KPPILLVAGK DDMEMCELNL EETGLTRKRG AEILPRQFEE
MYO6 Monoclonal antibody specifically detects MYO6 in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| MYO6 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| BC029719; DFNA22; DFNB37; KIAA0389; MYO6; myosin; myosin VI; myosin-VI; RGD1560646; Snell's waltzer; sv; tailchaser; Tlc; unconventional myosin; unconventional myosin-6; Unconventional myosin-VI | |
| Recombinant human MYO6 (immunogen range is 921-1260). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| MYO6 |
| K70041_12A6 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UM54 | |
| 4646 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.