missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYO7A Recombinant Rabbit Monoclonal Antibody (24GB665)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557096
This item is not returnable.
View return policy
Description
MYO7A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Myosin VIIa is a member of the myosin superfamily of actin-based motor proteins. Defects in the myosin VIIa gene are responsible for hearing impairment in shaker-1 (sh1) mice and causes Usher syndrome IB in humans. Usher syndrome associates congenital deafness, vestibular dysfunction, and retinitis pigmentosa and is the most common form of combined deafness and blindness. Structural features of myosin VIIa protein include an ATP binding N-terminal motor domain, a central region which possess five light-chain binding (IQ) motifs, and a C-terminal domain with three myosin tail homology (MyTH4) and talin-like homology regions.
Specifications
| MYO7A | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P97479, Q13402 | |
| MYO7A | |
| A synthesized peptide derived from human Myosin VIIa (aa 11-85). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB665 | |
| Unconjugated | |
| MYO7A | |
| DFNA11; DFNB2; Hdb; motor protein; MyO VIIa; Myo7; Myo7a; myosin; myosin 7a; myosin VIIA; myosin VIIA (Usher syndrome 1B (autosomal recessive, severe)); myosin-VIIa; Myosin-VIIa motor domain; MYOVIIA; MYU7A; nmf371; NSRD2; polka; sh1; sh-1; shaker 1; unconventional myosin-VIIa; USH1B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17921, 266714, 4647 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction