missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyoD Antibody (5.8A), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56511C
This item is not returnable.
View return policy
Description
MyoD Monoclonal specifically detects MyoD in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated, Knockdown Validated.
Specifications
| MyoD | |
| Monoclonal | |
| DyLight 650 | |
| P15172 | |
| MYOD1 | |
| The 5.8A antibody was made against recombinant mouse MyoD protein but it also recognizes human (myf3), rat, and cat homologs. The epitope of this antibody was mapped to a region within aa 180-189 of mouse MyoD (NP_002469). | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Growth and Development | |
| 4654 | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, KnockDown | |
| 5.8A | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated, Knockdown Validated | |
| bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| In Rh-30, a ∼45 kDa band should be observed. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction