missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MyoD Antibody (5.8A), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30942-25ug
This item is not returnable.
View return policy
Description
MyoD Monoclonal antibody specifically detects MyoD in Chicken,Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
Specifications
| MyoD | |
| Monoclonal | |
| Unconjugated | |
| Phosphate buffered saline | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Chicken, Human, Mouse, Rat | |
| Purified |
| ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence | |
| 5.8A | |
| ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation : 2 μg/mg of protein lysate, Immunohistochemistry-Paraffin : 2-4 μg/mL, Immunohistochemistry-Frozen : 2-4 μg/mL | |
| bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM | |
| Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with recombinant mouse MyoD protein and mouse myeloma Sp2/0-Ag14 cells. | |
| 25 μg | |
| Cancer, Cell Cycle and Replication, Growth and Development, Phospho Specific | |
| 4654 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction