Learn More
Novus Biologicals™ MyoD Antibody (5.8A), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-30942-100ug
 View more versions of this product
Product Code. 30067250
  • € 508.00 / 100µg
Description
MyoD Monoclonal antibody specifically detects MyoD in Chicken,Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MyoD
Monoclonal
Unconjugated
Phosphate buffered saline
Mouse
Protein A or G purified
RUO
Primary
Chicken, Human, Mouse, Rat
Purified
ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence
5.8A
ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation : 2 μg/mg of protein lysate, Immunohistochemistry-Paraffin : 2-4 μg/mL, Immunohistochemistry-Frozen : 2-4 μg/mL
bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM
Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with recombinant mouse MyoD protein and mouse myeloma Sp2/0-Ag14 cells.
100 μg
Cancer, Cell Cycle and Replication, Growth and Development, Phospho Specific
4654
Store at -20°C. Avoid freeze/thaw cycles.