missing translation for 'onlineSavingsMsg'
Learn More

MyoD Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-54153F

Product Code. 18513994

  • € 602.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MyoD Polyclonal antibody specifically detects MyoD in Human, Mouse samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MyoD
Polyclonal
PBS
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse
Purified
Immunofluorescence
FITC
bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM
A partial recombinant protein made to an N-terminal region of the human MyoD1 protein (within residues 1-150). [Swiss-Prot# P15172]
0.1 mL
Cancer, Cell Cycle and Replication, Growth and Development
4654
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.