missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyoD Rabbit anti-Human, Clone: MYOD1/3418R, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 528.00
Specifications
| Antigen | MyoD |
|---|---|
| Clone | MYOD1/3418R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
MyoD Monoclonal specifically detects MyoD in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| MyoD | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Growth and Development, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 4654 | |
| IgG | |
| Protein A purified |
| MYOD1/3418R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM | |
| Synthetic peptide corresponding to residues within (aa1-100) of human MyoD | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title