missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYOF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569432
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%; Zebrafish: 79%.
Mutations in dysferlin, a protein associated with the plasma membrane, can cause muscle weakness that affects both proximal and distal muscles. The protein encoded by this gene is a type II membrane protein that is structurally similar to dysferlin. It is a member of the ferlin family and associates with both plasma and nuclear membranes. The protein contains C2 domains that play a role in calcium-mediated membrane fusion events, suggesting that it may be involved in membrane regeneration and repair. Two transcript variants encoding different isoforms have been found for this gene. Other possible variants have been detected, but their full-length nature has not been determined.
Specifications
| MYOF | |
| Polyclonal | |
| Unconjugated | |
| Myof | |
| 2310004N10Rik; 2310051D19Rik; E030042N20Rik; FER1L3; fer-1-like 3, myoferlin; fer-1-like family member 3; fer-1-like protein 3; KIAA1207; Myof; Myoferlin; RGD1564216; zgc:63504; zgc:63504 protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26509 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NZM1 | |
| Myof | |
| synthetic peptide directed towards the C terminal region of human MYOF. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction