missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myogenin Antibody (MGN185 + F5D), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34684JF549
This item is not returnable.
View return policy
Description
Myogenin Monoclonal antibody specifically detects Myogenin in Human, Mouse, Rat, Porcine, Feline samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Myogenin | |
| Monoclonal | |
| Janelia Fluor 549 | |
| BHLHC3, bHLHc3Myogenic factor-4; myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4) | |
| Human Myogenin recombinant protein (MGN185); Rat Myogenin recombinant fragment containing amino acid 30-224 (F5D) (Uniprot: P15173) | |
| 0.1 mL | |
| Cancer, Stem Cell Markers, Transcription Factors and Regulators | |
| 4656 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MGN185 + F5D | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction