missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myogenin Mouse, Clone: F5D, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32986-0.02MG
This item is not returnable.
View return policy
Description
Myogenin Monoclonal specifically detects Myogenin in Human, Mouse, Rat, Porcine, Canine, Feline samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Myogenin | |
| Monoclonal | |
| 0.2mg/mL | |
| Western Blot 0.5-1.0ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml | |
| P15173 | |
| MYOG | |
| Rat myogenin peptide (aa 73-94) followed by rat myogenin recombinant fragment (aa30-224) (Epitope aa138-158) | |
| Protein A purified | |
| RUO | |
| Primary | |
| Myogenin is a member of the MyoD family of myogenic basic helix-loop-helix (bHLH) transcription factors that also includes MyoD, Myf-5, and MRF4 (also known as herculinor Myf-6). MyoD family members are expressed exclusively in skeletal muscle and play a key role in activating myogenesis by binding to enhancer sequences of muscle-specific genes. The regulatory domain of MyoD is approximately 70 amino acids in length and includes both a basic DNA binding motif and a bHLH dimerization motif. MyoD family members share about 80% amino acid homology in their bHLH motifs. Anti-myogenin labels the nuclei of myoblasts in developing muscle tissue, and is expressed in tumor cell nuclei of rhabdomyosarcoma and some leiomyosarcomas. Positive nuclear staining may occur in Wilms tumor. | |
| Store at 4C. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| F5D | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| BHLHC3, bHLHc3Myogenic factor-4; myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4) | |
| Mouse | |
| 34 kDa | |
| 0.02 mg | |
| Cancer, Stem Cell Markers, Transcription Factors and Regulators | |
| 4656 | |
| Human, Mouse, Rat, Pig, Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction