missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Myogenin Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519290
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, human, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
Myogenin is a muscle-specific transcription factor that can induce myogenesis in a variety of cell types in tissue culture. It is a member of a large family of proteins related by sequence homology, the helix-loop-helix proteins. It is essential for the development of functional skeletal muscle.
Specifications
| Myogenin | |
| Polyclonal | |
| Unconjugated | |
| MYOG | |
| bHLHc3; Class C basic helix-loop-helix protein 3; MYF4; Myf-4; myo; MYOD1-related protein; MYOG; Myogenic factor 4; Myogenic factor-4; myogenin; myogenin (myogenic factor 4) | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 17928 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P12979 | |
| MYOG | |
| Synthetic peptide sequence (TDAHNLHSLTSIVD) corresponding to the C-terminus amino acids of MYOG (aa 193-207). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction