Learn More
Invitrogen™ Myoglobin Monoclonal Antibody (7C3)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA122741
This item is not returnable.
View return policy
Description
MA1-22741 detects Myoglobin in Human samples.
This gene encodes a member of the globin superfamily and is expressed in skeletal and cardiac muscles. The encoded protein is a hemoprotein contributing to intracellular oxygen storage and transcellular facilitated diffusion of oxygen. At least three alternatively spliced transcript variants encoding the same protein have been reported.
Specifications
| Myoglobin | |
| Monoclonal | |
| 6.0 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P02144 | |
| MB | |
| Human cardiac myoglobin. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| 7C3 | |
| Unconjugated | |
| MB | |
| AI325109; Human myoglobin; Mb; MGC13548; myoglobgin; myoglobin; OTTHUMP00000197936; PVALB | |
| Mouse | |
| Protein A | |
| RUO | |
| 4151 | |
| 4°C | |
| Liquid |