missing translation for 'onlineSavingsMsg'
Learn More

Myomegalin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

€ 216.00 - € 588.00

Specifications

Antigen Myomegalin
Dilution Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:20-1:50
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18466610
View Documents
Novus Biologicals
NBP1-84428-25ul
25 μL
€ 216.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18016184
View Documents
Novus Biologicals
NBP1-84428
0.1 mL
€ 588.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Myomegalin Polyclonal specifically detects Myomegalin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Myomegalin
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
9659
This antibody was developed against Recombinant Protein corresponding to amino acids:DVLSSNEATMQSMESLLRAKGLEVEQLSTTCQNLQWLKEEMETKFSRWQKEQESIIQQLQTSLHDRNKEVEDLSATLLCKLGPGQSEIAEELCQRLQRKERMLQDLLSDRNKQVLEHEMEIQGLLQSVSTREQESQAAA
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:20-1:50
Polyclonal
Rabbit
Human, Mouse, Rat
Cardiomyopathy-associated protein 2, CMYA2, DKFZp781J054, KIAA0454, KIAA0477, MGC75440, MMGLcardiomyopathy associated 2, myomegalin, phosphodiesterase 4D interacting protein, Phosphodiesterase 4D-interacting protein
PDE4DIP
IgG
Affinity Purified
Specificity of human Myomegalin antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.