missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin 1B Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-97778
This item is not returnable.
View return policy
Description
Myosin 1B Polyclonal specifically detects Myosin 1B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Peptide ELISA.
Specifications
| Myosin 1B | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| MMIa, MMI-alpha, MYH-1c, MYO1B variant protein, Myosin I alpha, myosin IB, myosin-I alpha, myosin-Ib, myr1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_001123630.1; NP_036355.2). Reported variants represent identical protein: NP_001123630.1, NP_001155291.1. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Immunohistochemistry 5 - 10 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_001123630.1 | |
| MYO1B | |
| Peptide with sequence C-RTVEAKQEKVSTT corresponding to internal region according to NP_036355.2. | |
| 0.1 mg | |
| Vision | |
| 4430 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction