missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin 1E Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 196.00 - € 468.00
Specifications
| Antigen | Myosin 1E |
|---|---|
| Dilution | Western Blot 1:1000-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Myosin 1E Polyclonal antibody specifically detects Myosin 1E in Human samples. It is validated for Western BlotSpecifications
| Myosin 1E | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 4643 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| HuncM-IC, MGC104638, MYO1Cmyosin-IC, MYO1E variant protein, myosin IE, Myosin-Ic, myosin-Ie | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 809-1108 of human MYO1E (NP_004989.2). KEVLKRKIEIERILSVSLSTMQDDIFILHEQEYDSLLESVFKTEFLSLLAKRYEEKTQKQLPLKFSNTLELKLKKENWGPWSAGGSRQVQFHQGFGDLAVLKPSNKVLQVSIGPGLPKNSRPTRRNTTQNTGYSSGTQNANYPVRAAPPPPGYHQNGVIRNQYVPYPHAPGSQRSNQKSLYTSMARPPLPRQQSTSSDRVSQTPESLDFLKVPDQGAAGVRRQTTSRPPPAGGRPKPQPKPKPQVPQCKALYAYDAQDTDELSFNANDIIDIIKEDPSGWWTGRLRGKQGLFPNNYVTKI | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title