missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin 5a Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
€ 362.00
Specifications
| Antigen | Myosin 5a |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200-1:500, Knockdown Validated validation (Barthelemy-Requin et al). |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Myosin 5a Polyclonal specifically detects Myosin 5a in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| Myosin 5a | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 4644 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QNLQLPPEARIEASLQHEITRLTNENLDLMEQLEKQDKTVRKLKKQLKVFAKKIGELEVGQMENISPGQIIDEPIRPVNIPRKEKDFQGMLEYKKEDEQKLVKNLILELKPRGVAVNLIPG | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200-1:500, Knockdown Validated validation (Barthelemy-Requin et al). | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| Dilute myosin heavy chain, non-muscle, GS1myosin, heavy polypeptide kinase, MYH12myosin-12, MYO5, Myosin heavy chain 12, myosin V, myosin VA (heavy chain 12, myoxin), myosin VA (heavy polypeptide 12, myoxin), Myosin-12, myosin-Va, Myoxin, MYR12 | |
| MYO5A | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title