missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin heavy chain 11 Antibody (MYH11/2303R), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08486AF700
This item is not returnable.
View return policy
Description
Myosin heavy chain 11 Monoclonal antibody specifically detects Myosin heavy chain 11 in Human, Rat, Porcine, Bovine, Canine, Chicken, Feline, Guinea Pig, Rabbit samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Myosin heavy chain 11 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain | |
| Recombinant full-length human Myosin heavy chain 11 protein (Uniprot: P35749) | |
| 100 μg | |
| Hypoxia, Stem Cell Markers | |
| 4629 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| MYH11/2303R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat, Pig, Bovine, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction