missing translation for 'onlineSavingsMsg'
Learn More

Myosin heavy chain 11 Antibody (MYH11/8185R), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-24304MFV610

Product Code. 18516798

  • € 723.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Myosin heavy chain 11 Monoclonal antibody specifically detects Myosin heavy chain 11 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Myosin heavy chain 11
Monoclonal
mFluor Violet 610 SE
AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain
Recombinant fragment (around aa1-200) of human Myosin heavy chain 11 protein (exact sequence is proprietary)
0.1 mL
Hypoxia, Stem Cell Markers
4629
Store at 4°C in the dark.
IgG κ
Immunohistochemistry (Paraffin)
MYH11/8185R
50mM Sodium Borate
Rabbit
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.