missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin heavy chain 11 Antibody (MYH11/8185R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 268.00 - € 496.00
Specifications
| Antigen | Myosin heavy chain 11 |
|---|---|
| Clone | MYH11/8185R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18674015
|
Novus Biologicals
NBP3-23912-100ug |
100 μg |
€ 496.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18601084
|
Novus Biologicals
NBP3-23912-20ug |
20 μg |
€ 268.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myosin heavy chain 11 Monoclonal antibody specifically detects Myosin heavy chain 11 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Myosin heavy chain 11 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Hypoxia, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 4629 | |
| IgG κ | |
| Protein A or G purified |
| MYH11/8185R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain | |
| Recombinant fragment (around aa1-200) of human Myosin heavy chain 11 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title