missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin heavy chain 11 Antibody (MYH11/923 + SMMS-1), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47901V
This item is not returnable.
View return policy
Description
Myosin heavy chain 11 Monoclonal specifically detects Myosin heavy chain 11 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| Myosin heavy chain 11 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MYH11 | |
| Recombinant human Myosin heavy chain 11 protein (Myosin heavy chain 11/923); Human uterus extract (SMMS-1) (Uniprot: P35749) | |
| 0.1 mL | |
| Hypoxia, Stem Cell Markers | |
| Smooth muscle myosin heavy chain (SM-MHC) is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmentally regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2. The antibody may be useful for the study of breast tumors as the presence of an intact layer of myoepithelial cells is an important feature, which may distinguish benign breast lesions and carcinoma in situ from invasive tumors. | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| MYH11/923 + SMMS-1 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence | |
| AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction