missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin heavy chain 2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 214.00 - € 506.00
Specifications
| Antigen | Myosin heavy chain 2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625041
|
Novus Biologicals
NBP2-94624-0.02ml |
0.02 mL |
€ 214.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653391
|
Novus Biologicals
NBP2-94624-0.1ml |
0.1 mL |
€ 506.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myosin heavy chain 2 Polyclonal antibody specifically detects Myosin heavy chain 2 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Myosin heavy chain 2 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 4620 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| fast 2a myosin heavy chain, MYH2A, MYHas8, MyHC-2A, MyHC-IIa, MYHSA2, myosin heavy chain 2, myosin heavy chain 2a, myosin heavy chain IIa, myosin, heavy chain 2, skeletal muscle, adult, myosin, heavy polypeptide 2, skeletal muscle, adult, myosin-2, skeletal muscle, adult 2, type IIA myosin heavy chain | |
| A synthetic peptide corresponding to a sequence within amino acids 600-700 of human Myosin heavy chain 2 (NP_001093582.1). AQLFSGAQTAEGEGA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title