missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin Heavy Chain Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 57 publications
Brand: R&D Systems MAB4470
This item is not returnable.
View return policy
Description
Myosin Heavy Chain Monoclonal specifically detects Myosin Heavy Chain in Multi-Species samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| Myosin Heavy Chain | |
| Monoclonal | |
| LYOPH | |
| Western Blot, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| DFNA4, FLJ13881, FLJ43092, KIAA2034DKFZp667A1311, MHC16, MYH14 variant protein, MYH17, MYHC, myosin, Myosin heavy chain 14, Myosin heavy chain, non-muscle IIc, myosin, heavy chain 14, myosin, heavy chain 14, non-muscle, myosin, heavy polypeptide 14, myosin-14, NMHC II-C, NMHC-II-C, nonmuscle myosin heavy chain II-C, Non-muscle myosin heavy chain IIc | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 79784 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| MF20 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
| MYH14 | |
| Chicken pectoralis-derived Myosin | |
| 100 μg | |
| Primary | |
| Detects Myosin Heavy Chain in human, mouse, rat and other mammalian, avian, and amphibian species. | |
| Multi-species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction