missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin Light Chain 2 Antibody (3B2), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-30249AF700
This item is not returnable.
View return policy
Description
Myosin Light Chain 2 Monoclonal specifically detects Myosin Light Chain 2 in Human, Mouse, Xenopus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Myosin Light Chain 2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MYL2 | |
| Recombinant human MYL2 (1-166aa) purified from E. coli (NP_000423). | |
| 0.1 mL | |
| Phospho Specific | |
| 4633 | |
| Human, Mouse, Xenopus species | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 3B2 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| cardiac ventricular myosin light chain 2, CMH10myosin light chain 2, DKFZp779C0562, MLC2, MLC-2, MLC-2v, myosin regulatory light chain 2, ventricular/cardiac muscle isoform, myosin, light chain 2, regulatory, cardiac, slow, myosin, light polypeptide 2, regulatory, cardiac, slow, regulatory light chain of myosin, RLC of myosin, slow cardiac myosin regulatory light chain 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human MYL2 protein. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction