missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin light chain 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-88068-25ul
This item is not returnable.
View return policy
Description
Myosin light chain 3 Polyclonal specifically detects Myosin light chain 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Myosin light chain 3 | |
| Polyclonal | |
| Western Blot 1:100-1:500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Cardiac myosin light chain 1, CMH8Ventricular/slow twitch myosin alkali light chain, CMLC1, light polypeptide 3, alkali; ventricular, skeletal, slow, MLC1SBMyosin light chain 1, slow-twitch muscle B/ventricular isoform, MLC1V, myosin light chain 3, myosin, light chain 3, alkali; ventricular, skeletal, slow, VLC1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4634 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| MYL3 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KPEPKKDDAKAAPKAAPAPAPPPEPERPKEVEFDASKIKIEFTPEQIEEFKEAFMLFDRTPKCEM | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction