missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin Phosphatase [p Thr853] Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38643-20ul
This item is not returnable.
View return policy
Description
Myosin Phosphatase Polyclonal antibody specifically detects Myosin Phosphatase in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Myosin Phosphatase | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| MBSmyosin phosphatase, target subunit 1, MGC133042, Myosin phosphatase target subunit 1, Myosin phosphatase-targeting subunit 1, MYPT1protein phosphatase 1 regulatory subunit 12A, protein phosphatase 1, regulatory (inhibitor) subunit 12A, Protein phosphatase myosin-binding subunit | |
| A synthetic phosphorylated peptide around T853 of human Myosin Phosphatase (NP_002471.1).,, Sequence:, RSTGV | |
| 20 μL | |
| Phospho Specific, Protein Phosphatase | |
| 4659 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction